Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ SecureFit™ 400 Eye Protection

Low-profile, flexible temples are ideal for use with hearing protection. Contemporary, lightweight and wraparound lens design provides excellent coverage and good field of vision. Provides 99.9% UV protection.

$9.48

Specifications

Product Type Protective Eyewear
Disposable Reusable
View More Specs

Products 2
Catalog Number Mfr. No. For Use With (Application) Model Height (English) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Application) Model Height (English) Price Quantity & Availability  
03-052-855
View Documents
3M™
7100243523
Home Improvement, Building & Construction, Personal Safety, Eye Protectoin SF400C-WV-6-PS 8.5 in..
Each for $9.48
Only null left
Add to Cart
 
03-052-844
View Documents
3M™
7100112109
Home Improvement, Building & Construction, Personal Safety, Eye Protectoin SF400C-WV-6-PS 10.5 in. N/A N/A N/A
Description

Description

  • Lightweight, flat-temple, low-profile design works seamlessly with hearing protection
  • Wear all day - forget you have them on
  • Padded temples for added comfort over the ears
  • Soft, conformable nose pads provide customizable comfort
  • Lightweight, durable design for all-day comfort and lasting wear
  • Flexible temples conform comfortably to different head sizes
  • Anti-fog, scratch-resistant lenses help keep vision clear
  • Impact-resistant lenses block 99.9% of UV rays
  • Clear lenses help provide excellent visibility and can be used indoors or outdoors
  • Adjustable nose pads and soft-touch inlays increase comfort and provide grip
Specifications

Specifications

Protective Eyewear
Reusable
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.