Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ SecureFit™ 400 Series Readers Protective Eyewear

Features reading diopter to help workers who have difficulty reading small print or who are engaged in small detail work

$26.00 - $26.27

Specifications

Lens Coating Anti-fog
Lens Type Magnifying
Certifications/Compliance CSA Z94.3 and ANSI Z87.1-2010
Lens Tint Clear
Product Type Reader Protective Eyewear
View More Specs

Products 3
Catalog Number Mfr. No. Magnification Power Price Quantity & Availability  
Catalog Number Mfr. No. Magnification Power Price Quantity & Availability  
17-623-769
View Documents
3M™
7100115326
+1.50
Each for $26.27
Case of 20 EA for $525.31
Only null left
Add to Cart
 
17-623-770
View Documents
3M™
7100115325
+2.00
Each for $26.00
Case of 20 EA for $520.09
Only null left
Add to Cart
 
17-623-771
View Documents
3M™
7100115329
+2.50
Each for $26.27
Case of 20 EA for $525.31
Only null left
Add to Cart
 
Description

Description

Provides comfortable, durable, lightweight eye protection.

  • Clear anti-fog coated polycarbonate lens absorb 99.9% UV
  • Available with +1.5, +2.0, or +2.5 reading diopters
  • Pressure Diffusion Technology allows temples to naturally adjust to individual head sizes eliminating need to stock multiple frame sizes at worksites
  • Soft rubber nose bridge
  • Wide viewing area
  • Rubber temple tips
  • Color-accented temples
  • Frameless
  • Z87+ lens marking
  • One size fits all
  • 3M Catalog# SF415AF , 3M Catalog# SF420AF , 3M Catalog# SF425AF

Applications:

Facility Maintenance, Sawing, Masonary, Machining, Electrical, Machine Operations, Drilling, Assembly, Painting, Cleaning, Grinding, Pouring/Casting, Remediation, Sanding, Welding, Demolition, Riveting, Chipping, UV Protection, Power Fastening

Compliance

Meet ANSI Z87.1-2010 and CSA Z94.3 standards.

Specifications

Specifications

Anti-fog
CSA Z94.3 and ANSI Z87.1-2010
Reader Protective Eyewear
Magnifying
Clear
20/Cs.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.