Learn More
3M™ 3M™ SecureFit™ 500 Series Safety Glasses
3M™ SecureFit™ Protective Eyewear 500 Series features a stylized lens shape with temples that automatically self-adjust to help provide a secure and comfortable fit.
$225.06 - $250.12
Specifications
Product Type Protective Eyewear
For Use With (Application) Assembly, Machining, Remediation, Primary Metals, Power Fastening, Pouring/Casting, Pharmaceuticals, Painting, Oil and Gas, Mining, Military, Military Tactical, Military Maintenance, Repair and Operation (MRO), Masonary, Law Enforcement, Machine Operations, Automotive, Grinding, General Manufacturing, Food and Beverage Manufacturing, Food & Beverage, Facility Maintenance, Drilling, Demolition, Construction, Chiseling, Chemicals, Chemical Handling, Aviation, Sub Applications
Frame Color Clear
No. per Case 20/Cs.
Temple Style 3M Pressure Diffusion Temple Technology, Adjustable Temples
View More Specs
Products 4
Catalog Number Mfr. No. Lens Tint Lens Coating Price Quantity & Availability  
Catalog Number Mfr. No. Lens Tint Lens Coating Price Quantity & Availability  
19-072-253
View Documents
3M™
7100228722
Gray Anti-fog, Scratch-resistant
Case of 20 for $250.12
Only null left
Add to Cart
19-072-274
View Documents
3M™
7100228721
Gray I/O Anti-fog, Scratch-resistant
Case of 20 for $225.06
Only null left
Add to Cart
19-072-296
View Documents
3M™
7100228736
Clear Anti-fog, Scratch-resistant
Case of 20 for $225.06
Only null left
Add to cart
19-072-245
missing translation for 'viewDocuments'
3M™
7100228758
Clear N/A N/A N/A
Description
Description
  • Adjustable ratchet temples allow user to adjust lens for facial features and eyeglass position
  • Sporty design with contemporary lens shape
  • SecureFit™ Pressure Diffusion Temple technology design features slotted ribs that provide comfortable pressure equalization. The temples flex and self-adjust to the size of wearer's head.
  • Workers can feel confident wearing 3M eye protection that meets the requirements of ANSI Z87.1-2015 Standard as an Impact Rated Spectacle
  • Polycarbonate lenses absorb 99.9% of UVA and UVB rays.
Specifications
Specifications
Protective Eyewear
Clear
3M Pressure Diffusion Temple Technology, Adjustable Temples
Polycarbonate
Frameless
Assembly, Machining, Remediation, Primary Metals, Power Fastening, Pouring/Casting, Pharmaceuticals, Painting, Oil and Gas, Mining, Military, Military Tactical, Military Maintenance, Repair and Operation (MRO), Masonary, Law Enforcement, Machine Operations, Automotive, Grinding, General Manufacturing, Food and Beverage Manufacturing, Food & Beverage, Facility Maintenance, Drilling, Demolition, Construction, Chiseling, Chemicals, Chemical Handling, Aviation, Sub Applications
20/Cs.
Black
Plastic
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.