Learn More
3M™ Speedglas™ 9100 Welding Helmet
Catalog No. 19177851 Shop All 3M Products
Change view
Click to view available options
Product Type:
Inside protection plate
Sweatband
Welding Helmet
Quantity:
1 Ea.
10/Cs.
3/cs.
5/Cs.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
19-177-851 Welding Helmet 1 Ea.
19-177-853 Inside protection plate 10/Cs.
19-177-852 Inside protection plate 5/Cs.
19-177-854 Sweatband 3/cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-177-851 Supplier 3M™ Supplier No. 7010341605
Maintains personal protection with reduced weight and more comfort
  • Adjustable headband for excellent fit for each individual user
  • Easy back and forth adjustment of shield
  • Tiltable to the preferred angle
  • Adjustable delay with auto-darkening filter
  • 3M Catalog# 05-0013-41, 06-0200-30, 06-0200-52, 06-0200-54
TRUSTED_SUSTAINABILITY
Specifications
Model 05-0013-41
Product Type Welding Helmet
Quantity 1 Ea.