Learn More
3M™ Speedglas' FlexView' Helmet Replacement Kit
Catalog No. 191500514 Shop All 3M Products
Change view
Click to view available options
:
Helmet Replacement Kit; FlexView
Catalog No.
19-150-0514 Helmet Replacement Kit; FlexView
1 options
Catalog No. 19-150-0514 Supplier 3M™ Supplier No. 7000126787
Replacement kit includes front panel, welding filter holder, middle helmet panel, protection plate, and L and R (left and right) S stops. For 3M™ Speedglas™ FlexView System.
  • Air Duct
  • Replacement part
  • 3M Catalog# 04-0460-00
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) FlexView System
Includes Front panel, welding filter holder, middle helmet panel, protection plate and left and right “S” stops
Model 04-0460-00
Product Type Replacement FlexView Kit