Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Standard Chemical Sorbents

Catalog No. p-37658 Shop All 3M Products
Click to view available options
Capacity (English):
8 gal.
12 gal.
14 gal.
17 gal.
Type:
Chemical Sorbent Mini Boom
Chemical Sorbent Pad
Chemical Sorbent Particulate
Chemical Sorbent Pillow
Model:
P110
P200
P208
P300
P500
This item is not returnable. View return policy

Catalog No. 17-210-2

Supplier: 3M™ 7000001909

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Absorb virtually any liquid with these chemical sorbents

Meltblown polypropylene microfibers absorb liquids such as hydrofluoric acid and other aqueous-based chemicals, and soak up an average of 10 times their weight of most EPA-designated hazardous liquids.

  • Sorbents are dirt- and dust-free; contain no abrasive granules that can damage expensive equipment
  • Polypropylene base is chemically inert and incinerable; allows separation and recycling in many applications
  • Easy-to-spot yellow cartons protect sorbents from temperature and humidity for secure, long-term storage
  • Convenient dispenser boxes—suitable for tabletops or wall mounting—keep pads and pillows within easy reach Pads or rolls are ideal for use on work surfaces and can be trimmed to size
  • Mini booms surround and contain spills; pillows soak them up
  • 3M Catalog# P110, P200, P208, P300, P500

These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OSHA 29 CFR 1910.132
Color Yellow
Capacity (English) 12 gal.
Capacity (Metric) 45.4 L
Material Polypropylene
Type Chemical Sorbent Mini Boom
For Use With (Application) For control of spills of certain hazardous liquids
Model P200
Length (English) 4 ft.
Length (Metric) 1.21 m
Diameter (English) 3 in.
Diameter (Metric) 7.62 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.