Learn More
3M™ Tri-Flange™ Ear Plugs
Catalog No. 191300469 Shop All 3M Products
Click to view available options
Model:
P3000
P3001
Cord Material:
Cloth
Vinyl
Catalog No. 19-130-0469
Supplier: 3M™ 7000127187
Only null left
Add to Cart
Add to Cart
Tri-Flange™ flexible flanges adapt to the shape of the ear canal to comfortably reduce exposure to noise. The plastic stem allows the wearer to insert the ear plugs quickly and easily without touching the premolded tips.
  • Triple-flanged, premolded ear plugs fit comfortably in a wide range of sizes
  • Soft, flexible flanges conform to the shape of the ear canal
  • No need to roll ear plugs before fitting so tips stay clean even if hands are dirty
  • Washable and reusable
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
ANSI S3.1974 Yes
Cord Color Yellow
Cord Material Vinyl
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Assembly, Blasting, Cleaning, Composite Finishing, Demolition, Electrical, Facility Maintenance, Final Finish, Grinding, Machine Operations, Paint Preparation, Painting, Sanding, Welding
High Visibility Yes
Model P3000
Packaging Type Poly bag
Pairs per Package 400 Pairs/Cs
Plug Material ABS, elastomeric polymer
Plug Color Clear
Noise Reduction Rating (NRR) 26 dB
Corded Corded
Plug Shape Flanged
Disposable Reusable
Metal Detectable No
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.