Learn More
3M™ Versaflo™ M-400 Series PAPR Outer Shroud
Catalog No. 19039528 Shop All 3M Products
Click to view available options
Style:
Comfort
Flame Resistant
High Durability
Standard
This item is not returnable. View return policy
Catalog No. 19-039-528
Supplier: 3M™ 7100001830
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Outer shrouds are replacement parts for use with M-400 helmets.
  • The M-446 Outer Shroud is a comfortable, low-linting outer shroud for 3M™ Versaflo M-400 Helmets
  • Made from Nomex, the M-447 Outer Shroud is a flame resistant outer shroud for 3M™ Versaflo M-400 Helmets
  • Made from Cordura Nylon, the M-448 Outer Shroud is a high durability outer shroud for 3M™ Versaflo M-400 Helmets
  • 3M Catalog# M-445, M-446, M-447, M-448
TRUSTED_SUSTAINABILITY
Specifications
Assigned Protection Factor (APF) 1,000
Material Nomex
Product Type Outer Shroud
For Use With (Application) Cleaning, electrical, facility maintenance, painting, sanding, welding
For Use With (Equipment) M-400 helmet
Model M-447
Style Flame Resistant
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.