Description
- Has optional foam gasket attachment
- Foam gasket helps limit eye exposure to nuisance dust while providing additional cushioning
- Anti-Fog polycarbonate lens absorb 99.9%UV
- Assembly
- Cleaning
- Demolition
- Electrical
- Facility Maintenance
- Grinding
- Machine Operations
- Painting
- Remediation
- Sanding
- Welding
- 3M Catalog# 11872-00000-20 , 3M Catalog# 11873-00000-20 , 3M Catalog# 11874-00000-20
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3 standards.
Specifications
Specifications
| Lens Tint | Clear |
| For Use With (Application) | Assembly, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Remediation, Sanding, Welding |
| Includes | With optional foam gasket attachment |
| Model | 11872-00000-20 |
| Temple Style | Cord Control System Temple |
| Lens Material | Polycarbonate |
| Frame Material | Frameless |
CSA Z94.3-07, ANSI Z87.1-2010