Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3R,4R)-4-Acetoxy-3-[(R)-(tert-butyldimethylsilyloxy)ethyl]-2-azetidinone 98.0+%, TCI America™
SDP

Catalog No. A112425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A112425G 25 g
A11245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A112425G Supplier TCI America Supplier No. A112425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 76855-69-1
Molecular Formula C13H25NO4Si
Molecular Weight (g/mol) 287.43
MDL Number MFCD00077636,MFCD00077636
InChI Key GWHDKFODLYVMQG-UHFFFAOYNA-N
Synonym 2s,3r-3-1s-1-tert-butyldimethylsilyl oxy ethyl-4-oxoazetidin-2-yl acetate
PubChem CID 53860740
IUPAC Name 3-{1-[(tert-butyldimethylsilyl)oxy]ethyl}-4-oxoazetidin-2-yl acetate
SMILES CC(O[Si](C)(C)C(C)(C)C)C1C(NC1=O)OC(C)=O

Specifications

Melting Point 108°C
Color White
Quantity 25 g
UN Number 3077
Formula Weight 287.43
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material (3R,4R)-4-Acetoxy-3-[(R)-(tert-butyldimethylsilyloxy)ethyl]-2-azetidinone
EINECSNumber (5)-6118
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.