Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3R,4R)-(-)-D-Erythronolactone, 99+%, Thermo Scientific™

Catalog No. ac277172500
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15667-21-7
Molecular Formula C4H6O4
Molecular Weight (g/mol) 118.088
MDL Number MFCD00077763
InChI Key SGMJBNSHAZVGMC-PWNYCUMCSA-N
Synonym d-erythronolactone, 3r,4r-3,4-dihydroxydihydrofuran-2 3h-one, erythrono-1,4-lactone, 3r,4r-3,4-dihydroxyoxolan-2-one, d-erythrono-1,4-lactone, d-erythronic acid gamma-lactone, pubchem5782, 2r,3r-butane-2,3,4-triol-1,4 lactone, d-erythronic, a-lactone, erythronic acid g-lactone
PubChem CID 5325915
IUPAC Name (3R,4R)-3,4-dihydroxyoxolan-2-one
SMILES C1C(C(C(=O)O1)O)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 99°C to 102°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 250 mg
Specific Rotation Condition − 73.00 (20.00°C c=0.5,water)
Specific Rotation -71° to -75° (20°C, 589nm) (c=2, H2O)
Formula Weight 118.09
Percent Purity ≥99.0%
Physical Form Needles
Chemical Name or Material (3R, 4R)-(-)-D-Erythronolactone, 99.5+%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.