Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3S,5R)-5-(2,4-Difluorophenyl)-5-[(1H-1,2,4-triazol-1-yl)methyl]oxolan-3-ylmethyl p-Toluenesulfonate 98.0+%, TCI America™
SDP

Catalog No. D45371G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45371G 1 g
D4537200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45371G Supplier TCI America Supplier No. D45371G

Chemical Identifiers

CAS 149809-43-8
Molecular Formula C21H21F2N3O4S
Molecular Weight (g/mol) 449.473
MDL Number MFCD13195567
InChI Key DFWVLCJRFGIRAK-UHFFFAOYSA-N
Synonym p-Toluenesulfonic Acid (3S,5R)-5-(2,4-Difluorophenyl)-5-[(1H-1,2,4-triazol-1-yl)methyl]oxolan-3-ylmethyl Ester
PubChem CID 73090135
IUPAC Name [5-(2,4-difluorophenyl)-5-(1,2,4-triazol-1-ylmethyl)oxolan-3-yl]methyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCC2CC(OC2)(CN3C=NC=N3)C4=C(C=C(C=C4)F)F

Specifications

Melting Point 107°C
Color White-Yellow
Quantity 1 g
Formula Weight 449.47
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (3S,5R)-5-(2,4-Difluorophenyl)-5-[(1H-1,2,4-triazol-1-yl)methyl]oxolan-3-ylmethyl p-Toluenesulfonate
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.