Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(1,3-Oxazol-5-yl)benzenesulfonyl chloride, 97%, Thermo Scientific™

Catalog No. CC14503DA
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 337508-66-4
Molecular Formula C9H6ClNO3S
Molecular Weight (g/mol) 243.66
MDL Number MFCD02681962
InChI Key QCGUTLAPXWBIMM-UHFFFAOYSA-N
Synonym 4-1,3-oxazol-5-yl benzenesulfonyl chloride, 4-oxazol-5-yl benzene-1-sulfonyl chloride, 4-oxazol-5-yl-benzenesulfonyl chloride, benzenesulfonyl chloride, 4-5-oxazolyl, 4-1,3-oxazol-5-yl benzenesulfonylchloride, 4-1,3-oxazol-5-yl benzene-1-sulfonyl chloride, 4-1,3-oxazol-5-yl benzenesulphonyl chloride, 4-oxazol-5-ylbenzenesulfonyl-chloride, 4-oxazol-5-ylbenzene-sulfonyl-chloride, chloro 4-1,3-oxazol-5-yl phenyl sulfone
PubChem CID 2776415
IUPAC Name 4-(1,3-oxazol-5-yl)benzenesulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=C(C=C1)C1=CN=CO1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 243.67
Percent Purity 97%
Chemical Name or Material 4-(1, 3-Oxazol-5-yl)benzenesulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.