Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(1-Cyclohexen-1-yl)morpholine, 97%

Catalog No. aab2200414
Click to view available options
Quantity:
25 g
100 g

CAS: 670-80-4 | C10H17NO | 167.252 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 670-80-4
Molecular Formula C10H17NO
Molecular Weight (g/mol) 167.252
MDL Number MFCD00006163
InChI Key IIQFBBQJYPGOHJ-UHFFFAOYSA-N
Synonym 4-cyclohex-1-enyl-morpholine, 1-morpholino-1-cyclohexene, 1-morpholinocyclohexene, n-morpholino-1-cyclohexene, 4-1-cyclohexen-1-yl morpholine, morpholine, 4-1-cyclohexen-1-yl, 4-cyclohex-1-en-1-yl morpholine, 4-1-cyclohexenyl morpholine, 1-morpholin-1-ylcyclohexene, 1-n-morpholino cyclohexene
PubChem CID 69589
IUPAC Name 4-(cyclohexen-1-yl)morpholine
SMILES C1CCC(=CC1)N2CCOCC2

Specifications

Density 1.02
Boiling Point 118°C to 120°C (10 mmHg)
Flash Point 68°C (154°F)
Refractive Index 1.5146
Quantity 25 g
Beilstein 118696
Formula Weight 167.25
Percent Purity 97%
Chemical Name or Material 4-(1-Cyclohexen-1-yl)morpholine
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 211-579-6
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.