Learn More
4-(1-Piperazinyl)aniline, 95%
Description
4-Piperazinoaniline(CAS#67455-41-8)is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Piperazinoaniline(CAS#67455-41-8)is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Notes
Reactive with oxidizing agents. Keep at room temperature.

Specifications
Specifications
Melting Point | 119°C to 123°C |
Color | Purple to White |
Quantity | 1 g |
Formula Weight | 177.25 |
Percent Purity | 97% |
Physical Form | Powder |
Chemical Name or Material | 4-(1-Piperazinyl)aniline |
Safety and Handling
Hazard Category | H302-H315-H318-H335 |
Hazard Statement | GHS H Statement H301-H318-H315-H335 Toxic if swallowed. Causes serious eye damage. Causes skin irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.