Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-[(1-Pyrrolidinylsulfonyl)methyl]aniline 97.0+%, TCI America™
SDP

Catalog No. P21481G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21481G 1 g
P21485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P21481G Supplier TCI America Supplier No. P21481G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 334981-10-1
Molecular Formula C11H16N2O2S
Molecular Weight (g/mol) 240.32
MDL Number MFCD06795500
InChI Key VNSKHALYBQZMFW-UHFFFAOYSA-N
Synonym 1-[(4-Aminobenzyl)sulfonyl]pyrrolidine
PubChem CID 11184058
IUPAC Name 4-[(pyrrolidine-1-sulfonyl)methyl]aniline
SMILES NC1=CC=C(CS(=O)(=O)N2CCCC2)C=C1

Specifications

Melting Point 178°C
Color Yellow
Quantity 1 g
Formula Weight 240.32
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-[(1-Pyrrolidinylsulfonyl)methyl]aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.