Learn More
Promega Corporation 4-1BB Bioassay 5X
Catalog No. PRJA2355 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Druggable Target:
4-1BB (CD137/TNFRSF9)
Catalog No. Druggable Target
PRJA2355 4-1BB (CD137/TNFRSF9)
1 options
Catalog No. PRJA2355 Supplier Promega Corporation Supplier No. JA2355
Use a simpler, more consistent 4-1BB Bioassay for a mechanism-of-action (MOA)-based assay that is prequalified according to ICH guidelines. No primary cell culture required. Supplied in a convenient thaw-and-use format, also available in cell propagation model (CPM) format.
  • A simpler, more consistent 4-1BB bioassay.
  • Prequalified according to ICH guidelines.
  • Mechanism-of-action (MOA)-based assay.
  • No primary cell culture required.
  • Supplied in convenient thaw-and-use format.
  • Also available in cell propagation model (CPM) format.
  • High sensitivity and specificity.
  • Consistent and reproducible results.
  • Suitable for research and clinical applications.
  • Streamlined workflow reduces hands-on time.
Specifications
Detection Method Bioluminescence
Type Bioassay
Cell Line Jurkat
Druggable Target 4-1BB (CD137/TNFRSF9)