Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-1BB Ligand/TNFSF9 Antibody (2357A), HRP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB22952H
Description
4-1BB Ligand/TNFSF9 Monoclonal antibody specifically detects 4-1BB Ligand/TNFSF9 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| 4-1BB Ligand/TNFSF9 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rabbit | |
| 0.1 mL | |
| Apoptosis, Biologically Active Proteins | |
| 8744 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 2357A | |
| Flow Cytometry, CyTOF-ready | |
| 4-1BB ligand, 4-1BBL, 4-1BB-L, CD137L, homolog of mouse 4-1BB-L, receptor 4-1BB ligand, tumor necrosis factor (ligand) superfamily, member 9, tumor necrosis factor ligand superfamily member 9 | |
| E. coli-derived human 4-1BB Ligand/TNFSF9, Arg71-Glu254, Accession # P41273.1 | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction