Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-1BB Ligand/TNFSF9 Antibody (AT113-2), DyLight 594, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP311986DL594
Description
4-1BB Ligand/TNFSF9 Monoclonal antibody specifically detects 4-1BB Ligand/TNFSF9 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
4-1BB Ligand/TNFSF9 | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
IgG κ |
Western Blot, Flow Cytometry, Immunofluorescence | |
AT113-2 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
4-1BB ligand, 4-1BBL, 4-1BB-L, CD137L, homolog of mouse 4-1BB-L, receptor 4-1BB ligand, tumor necrosis factor (ligand) superfamily, member 9, tumor necrosis factor ligand superfamily member 9 | |
This antibody was raised by immunising LOU rats with soluble recombinant 4-1BB Ligand/TNFSF9:Fc fusion protein. | |
0.1 mL | |
Apoptosis, Biologically Active Proteins | |
8744 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction