Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-1BB Ligand/TNFSF9 Antibody (RM0067-3H19) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50 - $560.50
Specifications
Antigen | 4-1BB Ligand/TNFSF9 |
---|---|
Clone | RM0067-3H19 |
Concentration | LYOPH |
Dilution | Western Blot 1:500-1:1000, Flow Cytometry 1:50-1:200, CyTOF-ready |
Applications | Flow Cytometry, CyTOF, Western Blot |
Description
4-1BB Ligand/TNFSF9 Monoclonal specifically detects 4-1BB Ligand/TNFSF9 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
4-1BB Ligand/TNFSF9 | |
LYOPH | |
Flow Cytometry, CyTOF, Western Blot | |
Unconjugated | |
Rat | |
Apoptosis | |
P41274 | |
8744 | |
Mouse recombinant 4-1BB Ligand EC-domain | |
Primary | |
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant 4-1BB Ligand Extracellular domain. |
RM0067-3H19 | |
Western Blot 1:500-1:1000, Flow Cytometry 1:50-1:200, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
4-1BB ligand, 4-1BBL, 4-1BB-L, CD137L, homolog of mouse 4-1BB-L, receptor 4-1BB ligand, tumor necrosis factor (ligand) superfamily, member 9, tumor necrosis factor ligand superfamily member 9 | |
TNFSF9 | |
IgG2 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title