Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-1BB Ligand/TNFSF9 Antibody (RM0067-3H19), DyLight 755, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP121514IR
Description
4-1BB Ligand/TNFSF9 Monoclonal specifically detects 4-1BB Ligand/TNFSF9 in Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
4-1BB Ligand/TNFSF9 | |
Monoclonal | |
DyLight 755 | |
4-1BB ligand, 4-1BBL, 4-1BB-L, CD137L, homolog of mouse 4-1BB-L, receptor 4-1BB ligand, tumor necrosis factor (ligand) superfamily, member 9, tumor necrosis factor ligand superfamily member 9 | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant 4-1BB Ligand Extracellular domain. | |
Store at 4C in the dark. | |
IgG2 |
Western Blot, Flow Cytometry | |
RM0067-3H19 | |
Western Blot, Flow Cytometry | |
TNFSF9 | |
Mouse recombinant 4-1BB Ligand EC-domain | |
0.1 mL | |
Apoptosis | |
8744 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction