Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-1BB/TNFRSF9/CD137 Antibody (158332), DyLight 488, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB937K
This item is not returnable.
View return policy
Description
4-1BB/TNFRSF9/CD137 Monoclonal antibody specifically detects 4-1BB/TNFRSF9/CD137 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA (Matched Antibody Pair)Specifications
4-1BB/TNFRSF9/CD137 | |
Monoclonal | |
DyLight 488 | |
4-1BB, 4-1BB ligand receptor, CD137 antigen, CD137MGC2172, CDw137, FLJ43501, ILAhomolog of mouse 4-1BB, induced by lymphocyte activation (ILA), interleukin-activated receptor, homolog of mouse Ly63, receptor protein 4-1BB, T cell antigen ILA, T-cell antigen 4-1BB homolog, T-cell antigen ILA, tumor necrosis factor receptor superfamily member 9, tumor necrosis factor receptor superfamily, member 9 | |
Mouse myeloma cell line NS0-derived recombinant mouse 4-1BB, Val24-Leu187, Accession # P20334 | |
0.5 mL | |
Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Stem Cells | |
3604 | |
Store at 4°C in the dark. | |
IgG2a |
Western Blot, Flow Cytometry, ELISA | |
158332 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction