Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-1BB/TNFRSF9/CD137 Antibody (2356B), Allophycocyanin/Cy7, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB8381APCCY7
Description
4-1BB/TNFRSF9/CD137 Monoclonal antibody specifically detects 4-1BB/TNFRSF9/CD137 in Human samples. It is validated for Flow CytometrySpecifications
| 4-1BB/TNFRSF9/CD137 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 2356B | |
| Flow Cytometry | |
| 4-1BB, 4-1BB ligand receptor, CD137 antigen, CD137MGC2172, CDw137, FLJ43501, ILAhomolog of mouse 4-1BB, induced by lymphocyte activation (ILA), interleukin-activated receptor, homolog of mouse Ly63, receptor protein 4-1BB, T cell antigen ILA, T-cell antigen 4-1BB homolog, T-cell antigen ILA, tumor necrosis factor receptor superfamily member 9, tumor necrosis factor receptor superfamily, member 9 | |
| Chinese hamster ovary cell line CHO-derived recombinant human 4-1BB/TNFRSF9/CD137, Leu24-His183, Accession # Q07011 | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Stem Cells | |
| 3604 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction