Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-1BB/TNFRSF9/CD137 Antibody (LOB12.3) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP275200
Description
4-1BB/TNFRSF9/CD137 Monoclonal specifically detects 4-1BB/TNFRSF9/CD137 in Mouse samples. It is validated for Flow Cytometry, Functional.Specifications
4-1BB/TNFRSF9/CD137 | |
Monoclonal | |
Unconjugated | |
PBS with 0.02% Proclin 300 | |
TNFRSF9 | |
This antibody was generated by immunizing rats with recombinant CD137-Fc fusion protein. | |
0.2 mg | |
Primary | |
This antibody binds to mouse 4-1BB (CD137). | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
IgG2a κ |
Flow Cytometry, Functional Assay | |
LOB12.3 | |
Flow Cytometry 1:10 - 1:1000, Functional | |
4-1BB, 4-1BB ligand receptor, CD137 antigen, CD137MGC2172, CDw137, FLJ43501, ILAhomolog of mouse 4-1BB, induced by lymphocyte activation (ILA), interleukin-activated receptor, homolog of mouse Ly63, receptor protein 4-1BB, T cell antigen ILA, T-cell antigen 4-1BB homolog, T-cell antigen ILA, tumor necrosis factor receptor superfamily member 9, tumor necrosis factor receptor superfamily, member 9 | |
Rat | |
Protein A purified | |
Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Stem Cells | |
3604.0 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction