Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-1BB/TNFRSF9/CD137 Rabbit anti-Human, Clone: 4B4-1-1, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP3120290.025MG
Description
4-1BB/TNFRSF9/CD137 Monoclonal antibody specifically detects 4-1BB/TNFRSF9/CD137 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, NeutralizationSpecifications
4-1BB/TNFRSF9/CD137 | |
Monoclonal | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Block/Neutralize | |
4-1BB, 4-1BB ligand receptor, CD137 antigen, CD137MGC2172, CDw137, FLJ43501, ILAhomolog of mouse 4-1BB, induced by lymphocyte activation (ILA), interleukin-activated receptor, homolog of mouse Ly63, receptor protein 4-1BB, T cell antigen ILA, T-cell antigen 4-1BB homolog, T-cell antigen ILA, tumor necrosis factor receptor superfamily member 9, tumor necrosis factor receptor superfamily, member 9 | |
The original antibody was generated by immunizing BALB/c mice with a fusion protein of human 4-1BB/TNFRSF9/CD137 antigen and glutathione s transferase (GST). | |
0.025 mg | |
Apoptosis, Immune System Diseases, Immunology | |
3604 | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Neutralization | |
4B4-1-1 | |
Unconjugated | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction