Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(1H-Pyrazol-1-yl)benzaldehyde, ≥95%, Thermo Scientific™

Catalog No. p-2700452
Click to view available options
Quantity:
250 mg
1 g

Chemical Identifiers

CAS 99662-34-7
Molecular Formula C10H8N2O
Molecular Weight (g/mol) 172.187
MDL Number MFCD02681938
InChI Key PPGRDLZPSDHBIC-UHFFFAOYSA-N
Synonym 4-1h-pyrazol-1-yl benzaldehyde, 4-pyrazol-1-yl-benzaldehyde, 1-4'-formylphenyl pyrazole, 1-4-formylphenyl pyrazole, 4-1h-pyrazol-1-yl benzenecarbaldehyde, 4-pyrazol-1-yl benzaldehyde, 1-4-formylphenyl-1h-pyrazole, benzaldehyde, 4-1h-pyrazol-1-yl, 4-pyrazolylbenzaldehyde, pubchem22713
PubChem CID 2776477
IUPAC Name 4-pyrazol-1-ylbenzaldehyde
SMILES C1=CN(N=C1)C2=CC=C(C=C2)C=O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 172.19
Percent Purity 95%
Chemical Name or Material 4-(1H-Pyrazol-1-yl)benzaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.