Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Chloro-4-nitrophenyl)morpholine, 97%, Thermo Scientific™

Catalog No. AAH6306303
Click to view available options
Quantity:
250 mg
1 g
5 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55435-71-7
Molecular Formula C10H11ClN2O3
Molecular Weight (g/mol) 242.659
MDL Number MFCD00052898
InChI Key ZPKGWYXJULKUEH-UHFFFAOYSA-N
PubChem CID 108711
IUPAC Name 4-(2-chloro-4-nitrophenyl)morpholine
SMILES C1COCCN1C2=C(C=C(C=C2)[N+](=O)[O-])Cl

Specifications

Quantity 1 g
Synonym 4-2-chloro-4-nitrophenyl morpholine, morpholine, 4-2-chloro-4-nitrophenyl, n-2-chloro-4-nitrophenyl morpholine, 4-2-chloro-4-nitro-phenyl-morpholine, 3-chloro-4-morpholino-1-nitrobenzene, maybridge1_003410, acmc-1b11p, morpholine,4-2-chloro-4-nitrophenyl, 3-chloro-4-morpholin-4-yl-1-nitro-benzene
Formula Weight 242.66
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-(2-Chloro-4-nitrophenyl)morpholine
EINECSNumber 259-638-5
TSCA Yes
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.