Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Hydroxyethyl)morpholine 99.0+%, TCI America™
SDP

Catalog No. H0244100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0244100ML 100 mL
H024425ML 25 mL
H0244500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0244100ML Supplier TCI America Supplier No. H0244100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 622-40-2
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.175
MDL Number MFCD00006180
InChI Key KKFDCBRMNNSAAW-UHFFFAOYSA-N
Synonym 2-morpholinoethanol, 4-2-hydroxyethyl morpholine, 4-morpholineethanol, n-2-hydroxyethyl morpholine, 2-4-morpholinyl ethanol, morpholine ethanol, morpholineethanol, hydroxyethylmorpholine, ethanol, 2-morpholino, beta-morpholinoethanol
PubChem CID 61163
ChEBI CHEBI:67144
IUPAC Name 2-morpholin-4-ylethanol
SMILES C1COCCN1CCO

Specifications

Melting Point 2°C
Color Yellow
pH 10.7 (100g/L H2O 20°C)
Boiling Point 222°C
Quantity 100 mL
Formula Weight 131.18
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(2-Hydroxyethyl)morpholine
EINECSNumber (5)-6876
RTECSNumber QE3510000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.