Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Phenyleth-1-ynyl)thiophene-2-carbaldehyde, 97%, Thermo Scientific™

Catalog No. KM08089CB
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
KM08089CB 250 mg Amber Glass Bottle
KM08089DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. KM08089CB Supplier Thermo Scientific Chemicals Supplier No. KM08089CB
Add to Cart

Chemical Identifiers

CAS 175203-58-4
Molecular Formula C13H8OS
Molecular Weight (g/mol) 212.266
MDL Number MFCD00174359
InChI Key SAYKNBQCVYQQJF-UHFFFAOYSA-N
Synonym 4-2-phenyleth-1-ynyl thiophene-2-carbaldehyde, 4-2-phenylethynyl thiophene-2-carbaldehyde, 4-phenylethynyl-thiophene-2-carbaldehyde, 4-phenylethynyl thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde, 4-phenylethynyl, 4-phenylethynyl thiophene-2-carboxaldehyde
PubChem CID 2737139
IUPAC Name 4-(2-phenylethynyl)thiophene-2-carbaldehyde
SMILES C1=CC=C(C=C1)C#CC2=CSC(=C2)C=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 212.27
Percent Purity 97%
Chemical Name or Material 4-(2-Phenyleth-1-ynyl)thiophene-2-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.