Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(3,5-Dibromophenyl)-2,6-diphenylpyrimidine 98.0+%, TCI America™
SDP

Catalog No. D48951G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48951G 1 g
D4895200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48951G Supplier TCI America Supplier No. D48951G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 607740-08-9
Molecular Formula C22H14Br2N2
Molecular Weight (g/mol) 466.176
MDL Number MFCD16619323
InChI Key DLXBSWIBLRUGFU-UHFFFAOYSA-N
PubChem CID 23139721
IUPAC Name 4-(3,5-dibromophenyl)-2,6-diphenylpyrimidine
SMILES C1=CC=C(C=C1)C2=CC(=NC(=N2)C3=CC=CC=C3)C4=CC(=CC(=C4)Br)Br

Specifications

Melting Point 199°C
Color White
Quantity 1 g
Formula Weight 466.18
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-(3,5-Dibromophenyl)-2,6-diphenylpyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.