Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(3-Bromo-5-fluorobenzyl)morpholine, 96%, Thermo Scientific™

Catalog No. AAH6267303
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6267303 1 g
AAH62673MD 250 mg
AAH6267306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6267303 Supplier Thermo Scientific Chemicals Supplier No. H6267303

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 281652-25-3
Molecular Formula C11H13BrFNO
Molecular Weight (g/mol) 274.133
MDL Number MFCD01307672
InChI Key KRXZMNYOZJCBKI-UHFFFAOYSA-N
Synonym 4-3-bromo-4-fluorobenzyl morpholine, 4-3-bromo-4-fluorophenyl methyl morpholine, cambridge id 5424905, 2-fluoro-5-morpholin-4-ylmethyl-bromobenzene
PubChem CID 782859
IUPAC Name 4-[(3-bromo-4-fluorophenyl)methyl]morpholine
SMILES C1COCCN1CC2=CC(=C(C=C2)F)Br

Specifications

Quantity 1 g
Formula Weight 274.13
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 4-(3-Bromo-5-fluorobenzyl)morpholine
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.