Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(3-Bromophenyl)-2,6-diphenylpyrimidine 98.0+%, TCI America™
SDP

Catalog No. B49301G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49301G 1 g
B4930200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49301G Supplier TCI America Supplier No. B49301G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 864377-28-6
Molecular Formula C22H15BrN2
Molecular Weight (g/mol) 387.28
InChI Key BPMSGKUGXMWVBH-UHFFFAOYSA-N
PubChem CID 58943285
IUPAC Name 4-(3-bromophenyl)-2,6-diphenylpyrimidine
SMILES C1=CC=C(C=C1)C2=CC(=NC(=N2)C3=CC=CC=C3)C4=CC(=CC=C4)Br

Specifications

Melting Point 136°C
Color White
Quantity 1 g
Formula Weight 387.28
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-(3-Bromophenyl)-2,6-diphenylpyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.