Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzyl Alcohol 97.0+%, TCI America™
SDP

Catalog No. T28181G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28181G 1 g
T2818200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T28181G Supplier TCI America Supplier No. T28181G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 87736-88-7
Molecular Formula C9H7F3N2O
Molecular Weight (g/mol) 216.163
InChI Key UZHHTEASLDJYBO-UHFFFAOYSA-N
Synonym 3-[4-(Hydroxymethyl)phenyl]-3-(trifluoromethyl)-3H-diazirine
PubChem CID 11367905
IUPAC Name [4-[3-(trifluoromethyl)diazirin-3-yl]phenyl]methanol
SMILES C1=CC(=CC=C1CO)C2(N=N2)C(F)(F)F

Specifications

Color Yellow
Quantity 1 g
Formula Weight 216.16
Percent Purity ≥97.0% (HPLC)
Physical Form Liquid
Chemical Name or Material 4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzyl Alcohol
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.