Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4,4,4',4',4'-Hexafluoro-DL-valine 98.0+%, TCI America™
SDP

Catalog No. H1427200MG
Click to view available options
Quantity:
200 mg

Chemical Identifiers

CAS 16063-80-2
Molecular Formula C5H5F6NO2
Molecular Weight (g/mol) 225.09
MDL Number MFCD00014349
InChI Key KRNSHCKTGFAMPQ-UHFFFAOYSA-N
Synonym 2-Amino-4,4,4-trifluoro-3-(trifluoromethyl)butyric Acid
IUPAC Name 2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butanoic acid
SMILES NC(C(C(F)(F)F)C(F)(F)F)C(O)=O

Specifications

Color White
Quantity 200 mg
Formula Weight 225.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4,4,4′,4′,4′-Hexafluoro-DL-valine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.