Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4',4″-Trihydroxytriphenylmethane, 97%, Thermo Scientific™

Catalog No. p-4813035
Click to view available options
Quantity:
25 g
100 g

Chemical Identifiers

CAS 603-44-1
Molecular Formula C19H16O3
Molecular Weight (g/mol) 292.33
MDL Number MFCD00191589
InChI Key WFCQTAXSWSWIHS-UHFFFAOYSA-N
Synonym 4,4',4-methanetriyltriphenol, leucoaurin, 4,4',4-trihydroxytriphenylmethane, leucoaurine, leucaurin, phenol, methylidynetris, 4-bis 4-hydroxyphenyl methyl phenol, 4,4',4-methylidynetriphenol, 4,4',4-methylidynetrisphenol, phenol, 4,4',4-methylidynetris
PubChem CID 69047
IUPAC Name 4-[bis(4-hydroxyphenyl)methyl]phenol
SMILES OC1=CC=C(C=C1)C(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1

Specifications

Melting Point 244.0°C to 246.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 292.33
Percent Purity 97%
Chemical Name or Material 4, 4', 4''-Trihydroxytriphenylmethane
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 210-040-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.