Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Azoxydianisole 97.0+%, TCI America™
SDP

Catalog No. A055425G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 1562-94-3
Molecular Formula C14H14N2O3
Molecular Weight (g/mol) 258.277
MDL Number MFCD00008400
InChI Key KAEZRSFWWCTVNP-UHFFFAOYSA-N
Synonym 4,4'-azoxyanisole, p-azoxyanisole, 4,4'-azoxydianisole, p-azoxydianisole, p,p'-azoxyanisole, 4,4'-dimethoxyazoxybenzene, p,p'-azoxydianisole, paa liquid crystal, p,p'-dimethoxyazoxybenzene, azoxybenzene, 4,4'-dimethoxy
PubChem CID 15277
IUPAC Name (4-methoxyphenyl)-(4-methoxyphenyl)imino-oxidoazanium
SMILES COC1=CC=C(C=C1)N=[N+](C2=CC=C(C=C2)OC)[O-]

Specifications

Quantity 25 g
Formula Weight 258.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Azoxydianisole
EINECSNumber (3)-2828
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.