Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Biphenol, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
250g
Packaging:
Glass bottle
Plastic bottle
92-88-6
C12H10O2
186.21
MFCD00002348
VCCBEIPGXKNHFW-UHFFFAOYSA-N
4,4'-dihydroxybiphenyl,4,4'-biphenol,4,4'-biphenyldiol,biphenyl-4,4'-diol,1,1'-biphenyl-4,4'-diol,p,p'-biphenol,p,p'-diphenol,antioxidant dod,4,4'-dioxydiphenyl,p-dihydroxydiphenyl
7112
CHEBI:34367
4-(4-hydroxyphenyl)phenol
C1=CC(=CC=C1C2=CC=C(C=C2)O)O
92-88-6
C12H10O2
186.21
MFCD00002348
VCCBEIPGXKNHFW-UHFFFAOYSA-N
4,4'-dihydroxybiphenyl,4,4'-biphenol,4,4'-biphenyldiol,biphenyl-4,4'-diol,1,1'-biphenyl-4,4'-diol,p,p'-biphenol,p,p'-diphenol,antioxidant dod,4,4'-dioxydiphenyl,p-dihydroxydiphenyl
7112
CHEBI:34367
4-(4-hydroxyphenyl)phenol
C1=CC(=CC=C1C2=CC=C(C=C2)O)O
Color Beige to White or Gray
Melting Point 278°C to 283°C
Packaging Plastic bottle
Quantity 250g
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula HOC6H4C6H4OH
Beilstein 06, 991
Solubility Information Solubility in water: sparingly soluble
Formula Weight 186.21
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 4, 4'-Biphenol
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-200-5