Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Biphenyldicarboxylic acid, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
787-70-2
C14H10O4
242.23
MFCD00002554
NEQFBGHQPUXOFH-UHFFFAOYSA-N
biphenyl-4,4'-dicarboxylic acid,4,4'-biphenyldicarboxylic acid,1,1'-biphenyl-4,4'-dicarboxylic acid,4-4-carboxyphenyl benzoic acid,4,4'-bibenzoic acid,4,4'-diphenic acid,4,4'-dicarboxybiphenyl,p,p'-diphenic acid,usaf do-69,bibenzoic acid
13084
4-(4-carboxyphenyl)benzoic acid
C1=CC(=CC=C1C2=CC=C(C=C2)C(=O)O)C(=O)O
787-70-2
C14H10O4
242.23
MFCD00002554
NEQFBGHQPUXOFH-UHFFFAOYSA-N
biphenyl-4,4'-dicarboxylic acid,4,4'-biphenyldicarboxylic acid,1,1'-biphenyl-4,4'-dicarboxylic acid,4-4-carboxyphenyl benzoic acid,4,4'-bibenzoic acid,4,4'-diphenic acid,4,4'-dicarboxybiphenyl,p,p'-diphenic acid,usaf do-69,bibenzoic acid
13084
4-(4-carboxyphenyl)benzoic acid
C1=CC(=CC=C1C2=CC=C(C=C2)C(=O)O)C(=O)O
Melting Point 300.0°C
Quantity 5g
Color White to Beige
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (C6H4COOH)2
Formula Weight 242.23
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 4, 4'-Biphenyldicarboxylic acid
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.