Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Biphenyldisulfonic Acid 98.0+%, TCI America™
SDP

Catalog No. B166425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B166425G 25 g
B16645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B166425G Supplier TCI America Supplier No. B166425G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 5314-37-4
Molecular Formula C12H10NaO6S2
Molecular Weight (g/mol) 337.32
MDL Number MFCD00035762
InChI Key HGAPLVLNYHEDFN-UHFFFAOYSA-N
PubChem CID 79202
IUPAC Name sodium [1,1'-biphenyl]-4,4'-disulfonic acid
SMILES [Na+].OS(=O)(=O)C1=CC=C(C=C1)C1=CC=C(C=C1)S(O)(=O)=O

Specifications

Quantity 25 g
UN Number 2585
Formula Weight 314.33
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Biphenyldisulfonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.