Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Biphthalic Anhydride 98.0+%, TCI America™
SDP

Catalog No. B1326100G
Click to view available options
Quantity:
25 g
100 g

Chemical Identifiers

CAS 2420-87-3
Molecular Formula C16H6O6
Molecular Weight (g/mol) 294.218
MDL Number MFCD00039140
InChI Key WKDNYTOXBCRNPV-UHFFFAOYSA-N
PubChem CID 75494
IUPAC Name 5-(1,3-dioxo-2-benzofuran-5-yl)-2-benzofuran-1,3-dione
SMILES C1=CC2=C(C=C1C3=CC4=C(C=C3)C(=O)OC4=O)C(=O)OC2=O

Specifications

Melting Point 303°C
Color White-Yellow
Quantity 100 g
Synonym Diphenyl-3,3′,4,4′-tetracarboxylic Dianhydride
Formula Weight 294.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Biphthalic Anhydride
EINECSNumber (4)-0833
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.