Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Bis(4,6-diphenyl-1,3,5-triazin-2-yl)biphenyl (This product is unavailable in the U.S.) 95.0+%, TCI America™
SDP

Catalog No. B49771G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49771G 1 g
B4977200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49771G Supplier TCI America Supplier No. B49771G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 266349-83-1
Molecular Formula C42H28N6
Molecular Weight (g/mol) 616.73
MDL Number MFCD28291963
InChI Key UICMBMCOVLMLIE-UHFFFAOYSA-N
PubChem CID 22598582
IUPAC Name 2-[4'-(4,6-diphenyl-1,3,5-triazin-2-yl)-[1,1'-biphenyl]-4-yl]-4,6-diphenyl-1,3,5-triazine
SMILES C1=CC=C(C=C1)C1=NC(=NC(=N1)C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=NC(=NC(=N1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 1 g
Formula Weight 616.73
Percent Purity ≥95.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis(4,6-diphenyl-1,3,5-triazin-2-yl)biphenyl (This product is unavailable in the U.S.)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.