Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Bis(bromomethyl)biphenyl 95.0+%, TCI America™
SDP

Catalog No. B192125G
Click to view available options
Quantity:
1 g
5 g
25 g

Chemical Identifiers

CAS 20248-86-6
Molecular Formula C14H12Br2
Molecular Weight (g/mol) 340.06
MDL Number MFCD00094419
InChI Key HMUGRILXVBKBID-UHFFFAOYSA-N
PubChem CID 621238
IUPAC Name 4,4'-bis(bromomethyl)-1,1'-biphenyl
SMILES BrCC1=CC=C(C=C1)C1=CC=C(CBr)C=C1

Specifications

Melting Point 171°C
Quantity 25 g
UN Number 1759
Formula Weight 340.06
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis(bromomethyl)biphenyl
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.