Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Bis[di(3,5-xylyl)amino]-4″-phenyltriphenylamine 98.0+%, TCI America™
SDP

Catalog No. p-7119590 Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2269100MG 100 mg
B22691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2269100MG Supplier TCI America Supplier No. B2269100MG
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 249609-49-2
Molecular Formula C56H53N3
Molecular Weight (g/mol) 768.061
MDL Number MFCD03844780
InChI Key XPHQVAAGYDLYOC-UHFFFAOYSA-N
Synonym N-(4-Biphenylyl)-N,N-bis[4-[di(3,5-xylyl)amino]phenyl]amine
PubChem CID 22964147
IUPAC Name 4-N,4-N-bis(3,5-dimethylphenyl)-1-N-[4-(N-(3,5-dimethylphenyl)-3,5-dimethylanilino)phenyl]-1-N-(4-phenylphenyl)benzene-1,4-diamine
SMILES CC1=CC(=CC(=C1)N(C2=CC=C(C=C2)N(C3=CC=C(C=C3)C4=CC=CC=C4)C5=CC=C(C=C5)N(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C

Specifications

Quantity 100 mg
Formula Weight 768.06
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Bis[di(3,5-xylyl)amino]-4′′-phenyltriphenylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.