Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Chlorobenzoyl)piperidine, 97%, Thermo Scientific™

Catalog No. AAH6474003
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as pharmaceutical intermediate. The derivative of this compound, 2-amino-3-(4-chlorobenzoyl)-4-[4-(alkyl/aryl)piperazin-yl]thiophene are used as allosteric enhancers of the A1-adenosine receptor.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53220-41-0
Molecular Formula C12H14ClNO
Molecular Weight (g/mol) 223.70
MDL Number MFCD04113538
InChI Key IYGWDOXHCPQXKN-UHFFFAOYSA-N
Synonym 4-4-chlorobenzoyl piperidine, 4-chlorophenyl piperidin-4-yl methanone, 4-4-chlorophenyl carbonyl piperidine, 4-chlorophenyl-4-piperidinyl methanone, 4-chlorophenyl-piperidin-4-yl-methanone, maybridge1_003174, 4-chlorophenyl 4-piperidinyl methanonehydrochloride, 4-4-chlorobenzoyl-piperidine, 4-chlorophenyl 4-piperidyl ketone, 4-chlorophenyl piperidin-4-ylmethanone
PubChem CID 2777115
SMILES ClC1=CC=C(C=C1)C(=O)C1CCNCC1

Specifications

Melting Point 60°C to 63°C
Color Yellow
Quantity 1 g
Beilstein 158791
Solubility Information Slightly soluble in water.
IUPAC Name 4-(4-chlorobenzoyl)piperidine
Formula Weight 223.7
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-(4-Chlorobenzoyl)piperidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.