Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Di-n-octyloxyazoxybenzene 97.0+%, TCI America™
SDP

Catalog No. D11235G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 25729-12-8
Molecular Formula C28H42N2O3
Molecular Weight (g/mol) 454.655
MDL Number MFCD00059445
InChI Key LMDKWWQEAJSHLR-UHFFFAOYSA-N
PubChem CID 520227
IUPAC Name (4-octoxyphenyl)-(4-octoxyphenyl)imino-oxidoazanium
SMILES CCCCCCCCOC1=CC=C(C=C1)N=[N+](C2=CC=C(C=C2)OCCCCCCCC)[O-]

Specifications

Color Yellow
Quantity 5 g
Formula Weight 454.66
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Di-n-octyloxyazoxybenzene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.