Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4″-Diamino-p-terphenyl, 95%

Catalog No. aal0540303
Click to view available options
Quantity:
1 g

CAS: 3365-85-3 | C18H16N2 | 260.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3365-85-3
Molecular Formula C18H16N2
Molecular Weight (g/mol) 260.34
MDL Number MFCD00051532
InChI Key QBSMHWVGUPQNJJ-UHFFFAOYSA-N
Synonym 4,4-diamino-p-terphenyl, 4,4-diaminoterphenyl, 4-4-4-aminophenyl phenyl aniline, ccris 4965, p-terphenyl, 4,4-diamine, 1,1':4',1-terphenyl-4,4-diamine, 4'-4-aminophenyl-1,1'-biphenyl-4-amine, 4,4-diamino-4-terphenyl
PubChem CID 104949
IUPAC Name 4-[4-(4-aminophenyl)phenyl]aniline
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)N)C3=CC=C(C=C3)N

Specifications

Melting Point 244°C to 250°C
Quantity 1 g
Beilstein 2120295
Formula Weight 260.34
Percent Purity 95%
Chemical Name or Material 4,4''-Diamino-p-terphenyl
Hazard Category H302-H341
Hazard Statement GHS H Statement
H301-H341
Toxic if swallowed.
Suspected of causing genetic defects.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.