Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4,4-Diaminodiphenyl sulfide, ≥99.0% (GC), MilliporeSigma™ Supelco™
Click to view available options
Quantity:
250 mg
Description
Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications
Specifications
CAS | 139-65-1 |
Physical Form | Neat |
Linear Formula | S(C6H4 NH2)2 |
MDL Number | MFCD00025342 |
Melting Point | 105°C to 107°C |
Synonym | 4,4 ′-Thiodianiline |
Type | Analytical Standard |
Quantity | 250 mg |
Formula Weight | 216.30 |
Percent Purity | ≥99.0% (GC) |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction