Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Diethylaminophenylazo)pyridine 98.0+%, TCI America™
SDP

Catalog No. D25685G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25685G 5 g
D25681G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25685G Supplier TCI America Supplier No. D25685G

Chemical Identifiers

CAS 89762-42-5
Molecular Formula C15H18N4
Molecular Weight (g/mol) 254.337
MDL Number MFCD01631306
InChI Key FLPXBJDOCHBJCT-UHFFFAOYSA-N
PubChem CID 14671163
IUPAC Name N,N-diethyl-4-(pyridin-4-yldiazenyl)aniline
SMILES CCN(CC)C1=CC=C(C=C1)N=NC2=CC=NC=C2

Specifications

Quantity 5 g
Synonym 4-(4′-Pyridylazo)-N,N-diethylaniline
Formula Weight 254.34
Percent Purity ≥98.0% (T,HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Diethylaminophenylazo)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.