Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Difluorobenzhydrol, 98+%

Catalog No. p-7021858
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 365-24-2 | C13H10F2O | 220.219 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 365-24-2
Molecular Formula C13H10F2O
Molecular Weight (g/mol) 220.219
MDL Number MFCD00000357
InChI Key WCTZPQWLFWZYJE-UHFFFAOYSA-N
Synonym 4,4'-difluorobenzhydrol, bis 4-fluorophenyl methanol, bis-4-fluorophenyl-methanol, 4,4'-difluorobenzhydryl alcohol, bis 4-fluorophenyl methan-1-ol, 4,4'-diflurobenzylmethanol, pubchem14058, 4,4'-diflourobenzhydrol, 4,4'-difluoro benzhydrol, 4,4'-difluoro-benzhydrol
PubChem CID 123049
IUPAC Name bis(4-fluorophenyl)methanol
SMILES C1=CC(=CC=C1C(C2=CC=C(C=C2)F)O)F

Specifications

Melting Point 45°C to 49°C
Boiling Point 142°C to 144°C (3 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 2332258
Formula Weight 220.22
Percent Purity ≥98%
Chemical Name or Material 4,4'-Difluorobenzhydrol
EINECSNumber 206-671-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.