Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4,4'-Dihydroxybiphenyl, 99%

Click to view available options
Quantity:
25g
100g
500g
92-88-6
C12H10O2
186.21
MFCD00002348
VCCBEIPGXKNHFW-UHFFFAOYSA-N
4,4'-dihydroxybiphenyl,4,4'-biphenol,4,4'-biphenyldiol,biphenyl-4,4'-diol,1,1'-biphenyl-4,4'-diol,p,p'-biphenol,p,p'-diphenol,antioxidant dod,4,4'-dioxydiphenyl,p-dihydroxydiphenyl
7112
CHEBI:34367
4-(4-hydroxyphenyl)phenol
C1=CC(=CC=C1C2=CC=C(C=C2)O)O
92-88-6
C12H10O2
186.21
MFCD00002348
VCCBEIPGXKNHFW-UHFFFAOYSA-N
4,4'-dihydroxybiphenyl,4,4'-biphenol,4,4'-biphenyldiol,biphenyl-4,4'-diol,1,1'-biphenyl-4,4'-diol,p,p'-biphenol,p,p'-diphenol,antioxidant dod,4,4'-dioxydiphenyl,p-dihydroxydiphenyl
7112
CHEBI:34367
4-(4-hydroxyphenyl)phenol
C1=CC(=CC=C1C2=CC=C(C=C2)O)O
Applications
4,4′-Dihydroxybiphenyl is used to prepare plastics. It serves as a laboratory reagent and an intermediate in the preparation of polyether liquid crystals. Further, it is used as an antioxidant agent for rubber and latex. In addition to this, it is used in light-colored rubber products, food packaging and medical latex products.

Solubility
Slightly soluble in water.

Notes
Incompatible with oxidizing agents.
Quantity 500g
Odor Odorless
Melting Point 279°C to 285°C
Beilstein 1908886
Formula Weight 186.21
Percent Purity 99%
Chemical Name or Material 4,4'-Dihydroxybiphenyl

GHS H Statement
H312-H315-H319-H335
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

RTECSNumber : DV4725000

TSCA : TSCA